2-Ethylphenyl isothiocyanate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

008970-5G 008970-25G
FLUO008970-5GEA 0 GBP
FLUO008970-5G FLUO008970-25G
2-Ethylphenyl isothiocyanate
2-Ethylphenyl isothiocyanate
Formula: C₉H₉NS
Boiling Pt: 53 °C (0.1 mmHg)
Density: 1,076 g/cm³ (20 °C)
MDL Number: MFCD00041066
CAS Number: 19241-19-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR