2-Phenylethyl isothiocyanate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

009560-5G 009560-25G
FLUO009560-5GEA 0 GBP
FLUO009560-5G FLUO009560-25G
2-Phenylethyl isothiocyanate
2-Phenylethyl isothiocyanate
Formula: C₆H₅CH₂CH₂NCS
Density: 1.09 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004821
CAS Number: 2257-09-2
EINECS: 218-855-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR