Benzyl(dimethyl)amine, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
DMBA, N,N-Dimethylbenzylamine, N-Benzyldimethylamine, BDMA
M-Clarity™ quality level = MQ200
185582-100ML
185582-500ML
SIAL185582-100MLEA
64.5
GBP
SIAL185582-100ML
SIAL185582-500ML
Benzyl(dimethyl)amine, Sigma-Aldrich®
Benzyl(dimethyl)amine
Formula:
C₆H₅CH₂N(CH₃)₂ MW: 135.21 g/mol Boiling Pt: 183 Melting Pt: –75 °C Density: 0.915 g/cm³ (20 °C) Flash Pt: 53 Storage Temperature: Ambient |
MDL Number:
MFCD00008329 CAS Number: 103-83-3 EINECS: 203-149-1 UN: 2619 ADR: 8 II |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...