4'-Iodoacetophenone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 1-(4-Iodophenyl)ethan-1-one, p-Iodoacetophenone, 1-(4-Iodophenyl)ethanone

003065-25G 003065-100G
FLUO003065-25GEA 0 GBP
FLUO003065-25G FLUO003065-100G
4'-Iodoacetophenone
4'-Iodoacetophenone
Formula: C₈H₇IO
Boiling Pt: 145…150 °C (20 mmHg)
Melting Pt: 80…84 °C
Storage Temperature: Ambient
MDL Number: MFCD00045320
CAS Number: 13329-40-3
EINECS: 236-372-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR