3-(Trifluoromethyl)thiophenol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 3-Mercaptobenzotrifluoride, 3-(Trifluoromethyl)benzenethiol, 3-(Trifluoromethyl)benzene-1-thiol

002621-5G 002621-25G
FLUO002621-5GEA 0 GBP
FLUO002621-5G FLUO002621-25G
3-(Trifluoromethyl)thiophenol
3-(Trifluoromethyl)thiophenol
Formula: C₇H₅F₃S
Boiling Pt: 161…163 °C (1013 hPa)
MDL Number: MFCD00041142
CAS Number: 937-00-8
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR