α,α,α-Trifluoro-m-cresol
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
3-(Trifluoromethyl)phenol, 3-Hydroxybenzotrifluoride
001301-100G
001301-500G
001301-25G
FLUO001301-100GEA
0
GBP
FLUO001301-100G
FLUO001301-500G
FLUO001301-25G
α,α,α-Trifluoro-m-cresol
α,α,α-Trifluoro-m-cresol
Formula:
C₇H₅F₃O MW: 162.11 g/mol Boiling Pt: 178 Melting Pt: –2 °C Density: 1.34 g/cm³ (20 °C) Flash Pt: 73 Storage Temperature: Ambient |
MDL Number:
MFCD00002299 CAS Number: 98-17-9 EINECS: 202-645-5 UN: 3265 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...