α,α,α-Trifluoro-o-cresol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

001300-25G 001300-100G 001300-250G
FLUO001300-25GEA 0 GBP
FLUO001300-25G FLUO001300-100G FLUO001300-250G
α,α,α-Trifluoro-o-cresol
α,α,α-Trifluoro-o-cresol
Formula: C₇H₅F₃O
Boiling Pt: 147 °C (1013 hPa)
Melting Pt: 46 °C
Density: >1 g/cm³ (20 °C)
MDL Number: MFCD00002222
CAS Number: 444-30-4
EINECS: 207-148-7
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR