2-Fluorobenzyl alcohol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (2-Fluorophenyl)methanol, 2-Fluorobenzyl Alcohol

006196-25G 006196-100G
FLUO006196-25GEA 0 GBP
FLUO006196-25G FLUO006196-100G
2-Fluorobenzyl alcohol
2-Fluorobenzyl alcohol
Formula: C₇H₇FO
MW: 126.13 g/mol
Boiling Pt: 200
Density: 1.17 g/cm³ (20 °C)
MDL Number: MFCD00004601
CAS Number: 446-51-5
EINECS: 207-170-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR