2,4,6-Trifluorobenzyl alcohol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

007501-1G 007501-5G 007501-10G
FLUO007501-1GEA 0 GBP
FLUO007501-1G FLUO007501-5G FLUO007501-10G
2,4,6-Trifluorobenzyl alcohol
2,4,6-Trifluorobenzyl alcohol
Formula: C₇H₅F₃O
MDL Number: MFCD00061207
CAS Number: 118289-07-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR