2,4,5-Trifluorobenzyl alcohol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

007500-1G 007500-5G 007500-10G
FLUO007500-1GEA 0 GBP
FLUO007500-1G FLUO007500-5G FLUO007500-10G
2,4,5-Trifluorobenzyl alcohol
2,4,5-Trifluorobenzyl alcohol
Formula: C₇H₅F₃O
MDL Number: MFCD00061206
CAS Number: 144284-25-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR