2,3,4-Trifluorobenzyl alcohol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

007498-1G 007498-5G 007498-10G
FLUO007498-1GEA 0 GBP
FLUO007498-1G FLUO007498-5G FLUO007498-10G
2,3,4-Trifluorobenzyl alcohol
2,3,4-Trifluorobenzyl alcohol
Formula: C₇H₅F₃O
MW: 162.11 g/mol
MDL Number: MFCD00083551
CAS Number: 144284-24-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR