3,5-Difluorobenzyl alcohol
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
002114-25G
002114-100G
FLUO002114-25GEA
0
GBP
FLUO002114-25G
FLUO002114-100G
3,5-Difluorobenzyl alcohol
3,5-Difluorobenzyl alcohol
Formula:
C₇H₆F₂O Boiling Pt: 97…99 °C (9 mmHg) Density: 1,27 g/cm³ (20 °C) |
MDL Number:
MFCD00010313 CAS Number: 79538-20-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...