2,6-Difluorobenzyl alcohol
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
(2,6-Difluorophenyl)methanol
001961-25G
001961-100G
FLUO001961-25GEA
0
GBP
FLUO001961-25G
FLUO001961-100G
2,6-Difluorobenzyl alcohol
2,6-Difluorobenzyl alcohol
Formula:
C₇H₆F₂O Boiling Pt: 88 °C (14 mmHg) Density: 1,3 g/cm³ (20 °C) |
MDL Number:
MFCD00004603 CAS Number: 19064-18-7 EINECS: 242-792-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...