4-(Trifluoromethoxy)benzyl alcohol
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
002175-1G
002175-5G
002175-25G
FLUO002175-1GEA
0
GBP
FLUO002175-1G
FLUO002175-5G
FLUO002175-25G
4-(Trifluoromethoxy)benzyl alcohol
4-(Trifluoromethoxy)benzyl alcohol
Formula:
C₈H₇F₃O₂ Boiling Pt: 53…54 °C (0.5 mmHg) Density: 1,32 g/cm³ (20 °C) |
MDL Number:
MFCD00036029 CAS Number: 1736-74-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...