4-(Difluoromethoxy)benzyl alcohol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

007857-5G 007857-1G
FLUO007857-5GEA 0 GBP
FLUO007857-5G FLUO007857-1G
4-(Difluoromethoxy)benzyl alcohol
4-(Difluoromethoxy)benzyl alcohol
Formula: C₈H₈F₂O₂
MDL Number: MFCD00236228
CAS Number: 170924-50-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR