2,6-Difluoro-3-methylanisole

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

005745-1G 005745-5G
FLUO005745-1GEA 0 GBP
FLUO005745-1G FLUO005745-5G
2,6-Difluoro-3-methylanisole
2,6-Difluoro-3-methylanisole
Formula: C₈H₈F₂O
MW: 158.15 g/mol
MDL Number: MFCD01631341
CAS Number: 261763-33-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR