4-(Trifluoromethoxy)benzylamine
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
(4-(Trifluoromethoxy)phenyl)methanamine
001408-1G
001408-5G
001408-25G
001408-100G
FLUO001408-1GEA
0
GBP
FLUO001408-1G
FLUO001408-5G
FLUO001408-25G
FLUO001408-100G
4-(Trifluoromethoxy)benzylamine
4-(Trifluoromethoxy)benzylamine
Formula:
C₈H₈F₃NO Boiling Pt: 57 °C (1013 hPa) Density: 1,25 g/cm³ (20 °C) |
MDL Number:
MFCD00061237 CAS Number: 93919-56-3 EINECS: 300-040-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...