3-(Trifluoromethoxy)benzylamine
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
007551-1G
007551-5G
007551-10G
FLUO007551-1GEA
0
GBP
FLUO007551-1G
FLUO007551-5G
FLUO007551-10G
3-(Trifluoromethoxy)benzylamine
3-(Trifluoromethoxy)benzylamine
Formula:
C₈H₈F₃NO MW: 191.15 g/mol Boiling Pt: 211 °C (1013 hPa) Melting Pt: 55 °C |
MDL Number:
MFCD00061267 CAS Number: 93071-75-1 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...