4-Nitro-3-(trifluoromethyl)aniline
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
5-Amino-2-nitro-α,α,α-trifluorotoluene, 4-Nitro-α,α,α-trifluoro-m-toluidine, 5-Amino-2-nitrobenzotrifluoride
002683-25G
002683-100G
FLUO002683-25GEA
0
GBP
FLUO002683-25G
FLUO002683-100G
4-Nitro-3-(trifluoromethyl)aniline
4-Nitro-3-(trifluoromethyl)aniline
Formula:
C₇H₅F₃N₂O₂ MW: 206.12 g/mol Melting Pt: 129 Density: 1.752 g/cm³ (–123.15 °C) |
MDL Number:
MFCD00014717 CAS Number: 393-11-3 EINECS: 206-884-6 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...