4-Nitro-2-(trifluoromethyl)aniline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

002682-25G 002682-10G 002682-100G
FLUO002682-25GEA 0 GBP
FLUO002682-25G FLUO002682-10G FLUO002682-100G
4-Nitro-2-(trifluoromethyl)aniline
4-Nitro-2-(trifluoromethyl)aniline
Formula: C₇H₅F₃N₂O₂
Melting Pt: 94…95 °C
Density: 1.754 g/cm³ (–123.15 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007365
CAS Number: 121-01-7
EINECS: 204-443-2
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR