2-Nitro-4-(trifluoromethyl)aniline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Nitro-α,α,α-trifluoro-p-toluidine, 4-Amino-3-nitrobenzotrifluoride, 4-Amino-3-nitro-α,α,α-trifluorotoluene

003131-100G 003131-500G
FLUO003131-100GEA 0 GBP
FLUO003131-100G FLUO003131-500G
2-Nitro-4-(trifluoromethyl)aniline
2-Nitro-4-(trifluoromethyl)aniline
Formula: C₇H₅F₃N₂O₂
Melting Pt: 107 °C
MDL Number: MFCD00007155
CAS Number: 400-98-6
EINECS: 206-926-3
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR