4-Fluoro-m-toluidine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 4-Fluoro-3-methylaniline, 5-Amino-2-fluorotoluene

006273-5G 006273-25G 006273-100G
FLUO006273-5GEA 0 GBP
FLUO006273-5G FLUO006273-25G FLUO006273-100G
4-Fluoro-m-toluidine
4-Fluoro-m-toluidine
Formula: C₇H₈FN
MW: 125.15 g/mol
Boiling Pt: 85…86 °C (9 mmHg)
Melting Pt: 36 °C
Density: 1.12 g/cm³ (20 °C)
MDL Number: MFCD00025294
CAS Number: 452-69-7
EINECS: 207-207-7
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR