4-Fluoro-o-toluidine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

001687-25G 001687-100G 001687-10G
FLUO001687-25GEA 0 GBP
FLUO001687-25G FLUO001687-100G FLUO001687-10G
4-Fluoro-o-toluidine
4-Fluoro-o-toluidine
Formula: C₇H₈FN
Boiling Pt: 207 °C (1013 hPa)
Melting Pt: 14 °C
Density: 1.1263 g/cm³ (18 °C)
MDL Number: MFCD00007832
CAS Number: 452-71-1
EINECS: 207-208-2
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR