4-Fluoro-2-(trifluoromethyl)aniline
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
003127-100G
003127-25G
FLUO003127-100GEA
0
GBP
FLUO003127-100G
FLUO003127-25G
4-Fluoro-2-(trifluoromethyl)aniline
4-Fluoro-2-(trifluoromethyl)aniline
Formula:
C₇H₅F₄N Boiling Pt: 70 °C (1013 hPa) Density: 1.38 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00007831 CAS Number: 393-39-5 EINECS: 206-886-7 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...