5-Fluoro-o-toluidine
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
001692-25G
001692-100G
001692-500G
FLUO001692-25GEA
0
GBP
FLUO001692-25G
FLUO001692-100G
FLUO001692-500G
5-Fluoro-o-toluidine
5-Fluoro-o-toluidine
Formula:
C₇H₈FN Boiling Pt: 203 °C (1013 hPa) Melting Pt: 36…38 °C Density: 1.13 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007764 CAS Number: 367-29-3 EINECS: 206-689-6 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...