3-Fluoro-p-toluidine
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
001685-25G
001685-100G
FLUO001685-25GEA
0
GBP
FLUO001685-25G
FLUO001685-100G
3-Fluoro-p-toluidine
3-Fluoro-p-toluidine
Formula:
C₇H₈FN Boiling Pt: 97 °C (15 mmHg) Melting Pt: 37 °C Density: 1.09 g/cm³ (20 °C) |
MDL Number:
MFCD00007762 CAS Number: 452-77-7 EINECS: 207-212-4 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...