3-Fluoro-o-toluidine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

001693-5G 001693-25G 001693-100G
FLUO001693-5GEA 0 GBP
FLUO001693-5G FLUO001693-25G FLUO001693-100G
3-Fluoro-o-toluidine
3-Fluoro-o-toluidine
Formula: C₇H₈FN
Boiling Pt: 203 °C (1013 hPa)
Melting Pt: 7 °C
Density: 1.359 g/cm³ (20 °C)
MDL Number: MFCD00007760
CAS Number: 443-86-7
EINECS: 207-142-4
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR