α,α,α-Trifluoro-m-toluidine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

001602-500G 001602-1KG 001602-5KG
FLUO001602-500GEA 0 GBP
FLUO001602-500G FLUO001602-1KG FLUO001602-5KG
α,α,α-Trifluoro-m-toluidine
α,α,α-Trifluoro-m-toluidine
Formula: C₇H₆F₃N
Boiling Pt: 187 °C (1013 hPa)
Melting Pt: 5 °C
Density: 1.3 g/cm³ (20 °C)
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00007797
CAS Number: 98-16-8
EINECS: 202-643-4
UN: 2948
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR