2-Fluoro-6-(trifluoromethyl)aniline
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
001982-5G
001982-25G
FLUO001982-5GEA
0
GBP
FLUO001982-5G
FLUO001982-25G
2-Fluoro-6-(trifluoromethyl)aniline
2-Fluoro-6-(trifluoromethyl)aniline
Formula:
C₇H₅F₄N Boiling Pt: 155 °C (1013 hPa) Density: 1.388 g/cm³ (20 °C) |
MDL Number:
MFCD00061242 CAS Number: 144851-61-6 UN: 1992 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...