6-Fluoro-m-toluidine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

021823-50G 021823-10G
FLUO021823-50GEA 0 GBP
FLUO021823-50G FLUO021823-10G
6-Fluoro-m-toluidine
6-Fluoro-m-toluidine
Formula: C₇H₈FN
Boiling Pt: 88…90 °C (17 mmHg)
Density: 1.109 g/cm³ (20 °C)
MDL Number: MFCD00007654
CAS Number: 452-84-6
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR