2-Fluoro-5-(trifluoromethyl)aniline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

003129-25G 003129-100G 003129-500G
FLUO003129-25GEA 0 GBP
FLUO003129-25G FLUO003129-100G FLUO003129-500G
2-Fluoro-5-(trifluoromethyl)aniline
2-Fluoro-5-(trifluoromethyl)aniline
Formula: C₇H₅F₄N
Boiling Pt: 80…81 °C (26.7 hPa)
Density: 1.37 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007653
CAS Number: 535-52-4
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR