2-Fluoro-p-toluidine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Fluoro-4-methylaniline, 4-Amino-3-fluorotoluene

001686-5G 001686-25G 001686-100G
FLUO001686-5GEA 0 GBP
FLUO001686-5G FLUO001686-25G FLUO001686-100G
2-Fluoro-p-toluidine
2-Fluoro-p-toluidine
Formula: C₇H₈FN
Boiling Pt: 190 °C (1013 hPa)
Melting Pt: 3 °C
Density: 1.1084 g/cm³ (18 °C)
MDL Number: MFCD00040975
CAS Number: 452-80-2
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR