α,α,α-Trifluoro-o-toluidine
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
001984-500G
001984-1KG
001984-5KG
FLUO001984-500GEA
0
GBP
FLUO001984-500G
FLUO001984-1KG
FLUO001984-5KG
α,α,α-Trifluoro-o-toluidine
α,α,α-Trifluoro-o-toluidine
Formula:
C₇H₆F₃N Boiling Pt: 175 °C (1013 hPa) Melting Pt: –33 °C Density: 1.282 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007718 CAS Number: 88-17-5 EINECS: 201-806-7 UN: 2942 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...