4-Fluorobenzylamine
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
006199-50G
006199-250G
FLUO006199-50GEA
0
GBP
FLUO006199-50G
FLUO006199-250G
4-Fluorobenzylamine
4-Fluorobenzylamine
Formula:
C₇H₈FN Boiling Pt: 183 °C (1013 hPa) Density: 1.09 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00008120 CAS Number: 140-75-0 EINECS: 205-430-4 UN: 2735 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...