3-Fluorobenzylamine
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
006198-5G
006198-25G
FLUO006198-5GEA
0
GBP
FLUO006198-5G
FLUO006198-25G
3-Fluorobenzylamine
3-Fluorobenzylamine
Formula:
C₇H₈FN Boiling Pt: 87 °C (1013 hPa) Density: 1.09 g/cm³ (20 °C) |
MDL Number:
MFCD00008113 CAS Number: 100-82-3 EINECS: 202-891-3 UN: 2735 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...