3,5-Difluorobenzylamine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

004795-1G 004795-5G 004795-10G
FLUO004795-1GEA 0 GBP
FLUO004795-1G FLUO004795-5G FLUO004795-10G
3,5-Difluorobenzylamine
3,5-Difluorobenzylamine
Formula: C₇H₇F₂N
MW: 143.14 g/mol
Boiling Pt: 54 °C (1 mmHg)
Density: 1.21 g/cm³ (20 °C)
MDL Number: MFCD00061244
CAS Number: 90390-27-5
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR