2-Fluorobenzylamine
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
006197-25G
006197-100G
FLUO006197-25GEA
0
GBP
FLUO006197-25G
FLUO006197-100G
2-Fluorobenzylamine
2-Fluorobenzylamine
Formula:
C₇H₈FN Boiling Pt: 73 °C (1013 hPa) Density: 1.09 g/cm³ (20 °C) |
MDL Number:
MFCD00008107 CAS Number: 89-99-6 EINECS: 201-957-9 UN: 2735 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...