2,6-Difluorobenzylamine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

001963-25G 001963-5G
FLUO001963-25GEA 0 GBP
FLUO001963-25G FLUO001963-5G
2,6-Difluorobenzylamine
2,6-Difluorobenzylamine
Formula: C₇H₇F₂N
Boiling Pt: 174…177 °C (1013 hPa)
Density: 1.19 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010144
CAS Number: 69385-30-4
EINECS: 273-983-9
UN: 2735
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR