2,5-Difluorobenzylamine
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
004793-1G
004793-5G
FLUO004793-1GEA
0
GBP
FLUO004793-1G
FLUO004793-5G
2,5-Difluorobenzylamine
2,5-Difluorobenzylamine
Formula:
C₇H₇F₂N Boiling Pt: 76…78 °C (15 mmHg) Density: 1.22 g/cm³ (20 °C) |
MDL Number:
MFCD00010143 CAS Number: 85118-06-5 EINECS: 285-658-9 UN: 2735 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...