2,3,4-Trifluorobenzylamine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

007962-1G 007962-5G
FLUO007962-1GEA 0 GBP
FLUO007962-1G FLUO007962-5G
2,3,4-Trifluorobenzylamine
2,3,4-Trifluorobenzylamine
Formula: C₇H₆F₃N
MDL Number: MFCD00236313
CAS Number: 235088-67-2
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR