4-(Trifluoromethyl)phenyl isothiocyanate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

002620-5G 002620-25G
FLUO002620-5GEA 0 GBP
FLUO002620-5G FLUO002620-25G
4-(Trifluoromethyl)phenyl isothiocyanate
4-(Trifluoromethyl)phenyl isothiocyanate
Formula: C₈H₄F₃NS
Boiling Pt: 205…207 °C (1013 hPa)
Melting Pt: 29…31 °C
MDL Number: MFCD00039645
CAS Number: 1645-65-4
UN: 2923
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR