3-(Trifluoromethyl)phenyl isothiocyanate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: α,α,α-Trifluoro-m-tolyl isothiocyanate

002619-25G 002619-10G
FLUO002619-25GEA 0 GBP
FLUO002619-25G FLUO002619-10G
3-(Trifluoromethyl)phenyl isothiocyanate
3-(Trifluoromethyl)phenyl isothiocyanate
Formula: C₈H₄F₃NS
MW: 203.19 g/mol
Boiling Pt: 206…208 °C (1013 hPa)
Density: 1,33 g/cm³ (20 °C)
MDL Number: MFCD00004806
CAS Number: 1840-19-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR