3-(Trifluoromethyl)anthranilic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

005562-1G 005562-5G
FLUO005562-1GEA 0 GBP
FLUO005562-1G FLUO005562-5G
3-(Trifluoromethyl)anthranilic acid
3-(Trifluoromethyl)anthranilic acid
Formula: C₈H₆F₃NO₂
MDL Number: MFCD00274210
CAS Number: 313-12-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR