α,α,α,4-Tetrafluoro-m-toluic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CFF632.2.5 CFF632.5 CFF632.10 002222-5G 002222-25G 002222-1.5KG 002222-100G
CHMPCFF632.2.5EA 0 GBP
CHMPCFF632.2.5 CHMPCFF632.5 CHMPCFF632.10 FLUO002222-5G FLUO002222-25G FLUO002222-1.5KG FLUO002222-100G
α,α,α,4-Tetrafluoro-m-toluic acid
α,α,α,4-Tetrafluoro-m-toluic acid
Formula: C₈H₄F₄O₂
MDL Number: MFCD00061294
CAS Number: 67515-55-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR