α,α,α-Trifluoro-p-toluic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

002701-500G 002701-25G 002701-100G
FLUO002701-500GEA 0 GBP
FLUO002701-500G FLUO002701-25G FLUO002701-100G
α,α,α-Trifluoro-p-toluic acid
α,α,α-Trifluoro-p-toluic acid
Formula: C₈H₅F₃O₂
Melting Pt: 219…220 °C
MDL Number: MFCD00002562
CAS Number: 455-24-3
EINECS: 207-242-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR