5-Fluoro-o-toluic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

002249-1G 002249-5G 002249-25G
FLUO002249-1GEA 0 GBP
FLUO002249-1G FLUO002249-5G FLUO002249-25G
5-Fluoro-o-toluic acid
5-Fluoro-o-toluic acid
Formula: C₈H₇FO₂
MW: 154.14 g/mol
Boiling Pt: 263
Melting Pt: 134…135 °C
MDL Number: MFCD00042294
CAS Number: 33184-16-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR