3-Fluoro-p-toluic acid
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
001468-5G
001468-10G
001468-100G
001468-25G
FLUO001468-5GEA
0
GBP
FLUO001468-5G
FLUO001468-10G
FLUO001468-100G
FLUO001468-25G
3-Fluoro-p-toluic acid
3-Fluoro-p-toluic acid
Formula:
C₈H₇FO₂ Boiling Pt: 263 °C (1013 hPa) Melting Pt: 159…160 °C |
MDL Number:
MFCD00002490 CAS Number: 350-28-7 EINECS: 206-498-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...