α,α,α,3-Tetrafluoro-p-toluic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

007911-5G 007911-25G
FLUO007911-5GEA 0 GBP
FLUO007911-5G FLUO007911-25G
α,α,α,3-Tetrafluoro-p-toluic acid
α,α,α,3-Tetrafluoro-p-toluic acid
Formula: C₈H₄F₄O₂
MW: 208.11 g/mol
MDL Number: MFCD00236279
CAS Number: 115754-21-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR