α,α,α-Trifluoro-m-toluic acid
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
002133-25G
002133-100G
002133-250G
FLUO002133-25GEA
0
GBP
FLUO002133-25G
FLUO002133-100G
FLUO002133-250G
α,α,α-Trifluoro-m-toluic acid
α,α,α-Trifluoro-m-toluic acid
Formula:
C₈H₅F₃O₂ Boiling Pt: 238 °C (1013 hPa) Melting Pt: 106 °C Density: 1,371 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002519 CAS Number: 454-92-2 EINECS: 207-230-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...