6-Fluoro-m-toluic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

013846-25G 013846-5G
FLUO013846-25GEA 0 GBP
FLUO013846-25G FLUO013846-5G
6-Fluoro-m-toluic acid
6-Fluoro-m-toluic acid
Formula: C₈H₇FO₂
MW: 154.14 g/mol
MDL Number: MFCD00092819
CAS Number: 321-12-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR